missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 135°C to 140°C |
Color | Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 50 g |
Beilstein | 21, V,2, 409 |
Formula Weight | 279.69 |
Show More |
Safety and Handling
Product Identifier
- 4-¢4-Chloro-3-(trifluoromethyl)phenyl!-4-piperidinol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'