missing translation for 'onlineSavingsMsg'
Learn More

4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%, Thermo Scientific™

Product Code. 10328370
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 21928-50-7
Molecular Formula C12H13ClF3NO
Molecular Weight (g/mol) 279.69
MDL Number MFCD00066999
InChI Key RALRVIPTUXSBPO-UHFFFAOYSA-N
Synonym 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol
PubChem CID 89107
IUPAC Name 4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
SMILES C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 135°C to 140°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 21, V,2, 409
Formula Weight 279.69
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-¢4-Chloro-3-(trifluoromethyl)phenyl!-4-piperidinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'