missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%, Thermo Scientific™
£58.17 - £477.71
Chemical Identifiers
CAS | 21928-50-7 |
---|---|
Molecular Formula | C12H13ClF3NO |
Molecular Weight (g/mol) | 279.69 |
MDL Number | MFCD00066999 |
InChI Key | RALRVIPTUXSBPO-UHFFFAOYSA-N |
Synonym | 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol |
PubChem CID | 89107 |
IUPAC Name | 4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol |
SMILES | C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O |
Description
Chemical Identifiers
21928-50-7 | |
279.69 | |
RALRVIPTUXSBPO-UHFFFAOYSA-N | |
89107 | |
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O |
C12H13ClF3NO | |
MFCD00066999 | |
4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol | |
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol |
Specifications
21928-50-7 | |
100.0 | |
Beige | |
97% | |
MFCD00066999 | |
4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol | |
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O | |
279.69 | |
279.69 | |
Fine Crystalline Powder |
96.0 | |
135°C to 140°C | |
Authentic | |
C12H13ClF3NO | |
21, V,2, 409 | |
RALRVIPTUXSBPO-UHFFFAOYSA-N | |
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol | |
89107 | |
97% | |
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 244-665-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-¢4-Chloro-3-(trifluoromethyl)phenyl!-4-piperidinol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only