missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenoxy)piperidine,98%, Thermo Scientific Chemicals

£68.80 - £239.00

Chemical Identifiers

CAS 97839-99-1
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD04113544
InChI Key VOQMPZXAFLPTMM-UHFFFAOYSA-N
PubChem CID 5199889
IUPAC Name 4-(4-chlorophenoxy)piperidine
SMILES ClC1=CC=C(OC2CCNCC2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11499680
View Documents
Thermo Scientific Alfa Aesar
H50944.MD
250 mg
£68.80
250mg
Please to purchase this item. Need a web account? Register with us today!
11409690
View Documents
Thermo Scientific Alfa Aesar
H50944.03
1 g
£239.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

97839-99-1
211.69
VOQMPZXAFLPTMM-UHFFFAOYSA-N
4-(4-chlorophenoxy)piperidine
C11H14ClNO
MFCD04113544
5199889
ClC1=CC=C(OC2CCNCC2)C=C1

Specifications

97839-99-1
MFCD04113544
Slightly soluble in water.
ClC1=CC=C(OC2CCNCC2)C=C1
211.69
211.69
4-(4-Chlorophenoxy)piperidine
C11H14ClNO
4-4-chlorophenoxy piperidine, 4-4-chloro-phenoxy-piperidine, piperidine, 4-4-chlorophenoxy, acmc-20amwb, 4-p-chlorophenoxy piperidine, 4-4-chlorophenyl oxy piperidine, 4-chloro-1-4-piperidyloxy benzene, piperidine,4-4-chlorophenoxy, piperidine, 4-4-chlorophenoxy-, hydrochloride
VOQMPZXAFLPTMM-UHFFFAOYSA-N
4-(4-chlorophenoxy)piperidine
5199889
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only