missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline, Thermo Scientific Chemicals

Product Code. 10309020
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39512-49-7
Molecular Formula C11H15Cl2NO
Molecular Weight (g/mol) 248.15
MDL Number MFCD00006001
InChI Key PXAJMBHRLGKPQV-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine
PubChem CID 38282
IUPAC Name 4-(4-chlorophenyl)piperidin-4-ol
SMILES [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1

Specifications

Absorbance (1 w/v% soln. in DMF) (1 cm silica cell), (1 w/v% soln. in DMF) (1 cm silica cell) at 0nm, 0.05 max. at 440nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point 139°C to 141°C
Color Cream-White to White
Loss on Drying 0.5% max. (1 g, 105°C, 2 hrs, 760mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 340mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,acetone and dichloromethane
Formula Weight 211.69
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)-4-hydroxypiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4-chlorophenyl)piperidin-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'