missing translation for 'onlineSavingsMsg'
Learn More
4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™ Product Code.: 10049491

4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10049491
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10049491

Brand: Thermo Scientific Maybridge SB02037DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 95233-37-7
Molecular Formula C13H15ClO2
Molecular Weight (g/mol) 238.711
MDL Number MFCD06797626
InChI Key NXXDIEYTMQYWJU-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
PubChem CID 2735792
IUPAC Name 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula ClC6H4C6H10CO2H
Formula Weight 238.71
Percent Purity 97%
Chemical Name or Material (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.