Learn More
4-(4-chlorophenyl)piperidin-4-ol, 97%, Thermo Scientific™
£54.15 - £173.00
Chemical Identifiers
CAS | 39512-49-7 |
---|---|
Molecular Formula | C11H15Cl2NO |
Molecular Weight (g/mol) | 248.15 |
MDL Number | MFCD00006001 |
InChI Key | PXAJMBHRLGKPQV-UHFFFAOYSA-N |
Synonym | 4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine |
PubChem CID | 38282 |
IUPAC Name | 4-(4-chlorophenyl)piperidin-4-ol |
SMILES | [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10143014
|
Thermo Scientific Maybridge
BTB09284EA |
10 g |
£54.15
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10396784
|
Thermo Scientific Maybridge
BTB09284EB |
25 g |
£116.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10566615
|
Thermo Scientific Maybridge
BTB09284FA |
100 g |
£173.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
39512-49-7 | |
248.15 | |
PXAJMBHRLGKPQV-UHFFFAOYSA-N | |
38282 | |
[H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1 |
C11H15Cl2NO | |
MFCD00006001 | |
4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine | |
4-(4-chlorophenyl)piperidin-4-ol |
Specifications
39512-49-7 | |
100.0 | |
Amber glass bottle | |
MFCD00006001 | |
PXAJMBHRLGKPQV-UHFFFAOYSA-N | |
4-(4-chlorophenyl)piperidin-4-ol | |
38282 | |
97% |
96.5 | |
97% | |
C11H15Cl2NO | |
4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine | |
[H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1 | |
248.15 | |
211.69 | |
4-(4-Chlorophenyl)piperidin-4-ol |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.