missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-chlorophenyl)pyrimidine-2-thiol, 97%, Thermo Scientific™
£18.40 - £133.00
Chemical Identifiers
CAS | 175203-08-4 |
---|---|
Molecular Formula | C10H7ClN2S |
Molecular Weight (g/mol) | 222.69 |
MDL Number | MFCD00174262 |
InChI Key | TWKIWOOKHPWUIM-UHFFFAOYSA-N |
Synonym | 4-4-chlorophenyl pyrimidine-2-thiol, 4-4-chlorophenyl-2-pyrimidinethiol, 4-4-chloro-phenyl-pyrimidine-2-thiol, 6-4-chlorophenyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-4-chlorophenyl, maybridge1_004888, 6-4-chlorophenyl pyrimidine-2 1h-thione, 4-4-chlorophenyl-3h-pyrimidine-2-thione |
PubChem CID | 2735805 |
IUPAC Name | 6-(4-chlorophenyl)-1H-pyrimidine-2-thione |
SMILES | ClC1=CC=C(C=C1)C1=CC=NC(=S)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10723633
|
Thermo Scientific Maybridge
KM05717DA |
1 g | Amber Glass Bottle |
£18.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10232552
|
Thermo Scientific Maybridge
KM05717EA |
10 g | Amber Glass Bottle |
£133.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175203-08-4 | |
222.69 | |
TWKIWOOKHPWUIM-UHFFFAOYSA-N | |
2735805 | |
ClC1=CC=C(C=C1)C1=CC=NC(=S)N1 |
C10H7ClN2S | |
MFCD00174262 | |
4-4-chlorophenyl pyrimidine-2-thiol, 4-4-chlorophenyl-2-pyrimidinethiol, 4-4-chloro-phenyl-pyrimidine-2-thiol, 6-4-chlorophenyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-4-chlorophenyl, maybridge1_004888, 6-4-chlorophenyl pyrimidine-2 1h-thione, 4-4-chlorophenyl-3h-pyrimidine-2-thione | |
6-(4-chlorophenyl)-1H-pyrimidine-2-thione |
Specifications
175203-08-4 | |
97% | |
MFCD00174262 | |
TWKIWOOKHPWUIM-UHFFFAOYSA-N | |
6-(4-chlorophenyl)-1H-pyrimidine-2-thione | |
2735805 | |
97% |
97.0 | |
C10H7ClN2S | |
4-4-chlorophenyl pyrimidine-2-thiol, 4-4-chlorophenyl-2-pyrimidinethiol, 4-4-chloro-phenyl-pyrimidine-2-thiol, 6-4-chlorophenyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-4-chlorophenyl, maybridge1_004888, 6-4-chlorophenyl pyrimidine-2 1h-thione, 4-4-chlorophenyl-3h-pyrimidine-2-thione | |
ClC1=CC=C(C=C1)C1=CC=NC(=S)N1 | |
222.69 | |
222.7 | |
4-(4-Chlorophenyl)pyrimidine-2-thiol |