Learn More
4,4'-Di-n-nonyl-2,2'-bipyridine, 97%, Thermo Scientific Chemicals
Description
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 61°C to 63°C |
Quantity | 1 g |
Formula Weight | 408.67 |
Percent Purity | 97% |
Chemical Name or Material | 4,4'-Di-n-nonyl-2,2'-bipyridine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.