missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminoazobenzene, 95%

Product Code. 11342286
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11342286

Brand: Thermo Scientific Alfa Aesar L03743.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 538-41-0 | C12H12N4 | 212.26 g/mol

4,4'-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

Solubility
Very soluble in ethanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-41-0
Molecular Formula C12H12N4
Molecular Weight (g/mol) 212.26
MDL Number MFCD00041892
InChI Key KQIKKETXZQDHGE-UHFFFAOYSA-N
Synonym 4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
PubChem CID 10855
SMILES NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1

Specifications

Melting Point ∽245°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 745553
Merck Index 14,2977
Solubility Information Very soluble in ethanol. Insoluble in water.
IUPAC Name 4-[2-(4-aminophenyl)diazen-1-yl]aniline
Formula Weight 212.26
Percent Purity 95%
Chemical Name or Material 4,4'-Diaminoazobenzene
compliance-icons
Product Identifier
  • 4,4'-Diaminoazobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.