missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dichlorobutyrophenone, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 29°C to 30°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | ClC6H4CO(CH2)3Cl |
Quantity | 25 g |
Solubility Information | Solubility in water: insoluble |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'