missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzhydrol, 98+%, Thermo Scientific Chemicals

Artikelnummer. 11432038
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 728-87-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008410
InChI Key ZODAOVNETBTTJX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh
PubChem CID 69768
IUPAC Name bis(4-methoxyphenyl)methanol
SMILES COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1

Spezifikation

Melting Point 69°C to 71°C
Quantity 10 g
Beilstein 748532
Solubility Information Soluble in water (0.44g/L @ 25°C),methanol (0.1g/mL).
Formula Weight 244.29
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethoxybenzhydrol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'