Learn More
4,4-Dimethyl-2-cyclohexen-1-one, 97%, Thermo Scientific Chemicals
£56.60 - £213.00
Chemical Identifiers
CAS | 1073-13-8 |
---|---|
Molecular Formula | C8H12O |
Molecular Weight (g/mol) | 124.18 |
MDL Number | MFCD00009695 |
InChI Key | HAUNPYVLVAIUOO-UHFFFAOYSA-N |
Synonym | 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one |
PubChem CID | 136839 |
IUPAC Name | 4,4-dimethylcyclohex-2-en-1-one |
SMILES | CC1(C)CCC(=O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10471501
|
Thermo Scientific Acros
319540050 |
5 g | Glass bottle |
£56.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697294
|
Thermo Scientific Acros
319540250 |
25 g | Glass bottle |
£213.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1073-13-8 | |
124.18 | |
HAUNPYVLVAIUOO-UHFFFAOYSA-N | |
136839 | |
CC1(C)CCC(=O)C=C1 |
C8H12O | |
MFCD00009695 | |
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one | |
4,4-dimethylcyclohex-2-en-1-one |
Specifications
1073-13-8 | |
100.0 | |
0.9400g/mL | |
64°C | |
96% min. (GC) | |
(CH3)2C6H6(=O) | |
MFCD00009695 | |
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one | |
CC1(C)CCC(=O)C=C1 | |
124.18 | |
124.18 | |
Liquid |
96.0 | |
Brown | |
72.0°C to 73.5°C (20.0 mmHg) | |
Authentic | |
C8H12O | |
1.4710 to 1.4740 | |
0.94 | |
HAUNPYVLVAIUOO-UHFFFAOYSA-N | |
4,4-dimethylcyclohex-2-en-1-one | |
136839 | |
97% | |
4, 4-Dimethyl-2-cyclohexen-1-one |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO – Research Use Only