missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-oxazoline, 98%, Thermo Scientific Chemicals
£82.10 - £348.00
Chemical Identifiers
CAS | 30093-99-3 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.13 |
MDL Number | MFCD00014507 |
InChI Key | KOAMXHRRVFDWRQ-UHFFFAOYSA-N |
Synonym | 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl |
PubChem CID | 121630 |
SMILES | CC1(C)COC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10313872
|
Thermo Scientific Acros
355270100 |
10 g | Glass bottle |
£82.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11976821
|
Thermo Scientific Acros
355270500 |
50 g | Glass bottle |
£348.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30093-99-3 | |
99.13 | |
KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
121630 |
C5H9NO | |
MFCD00014507 | |
4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
CC1(C)COC=N1 |
Specifications
30093-99-3 | |
100.0 | |
0.9400g/mL | |
1°C | |
97.5% min. (GC) | |
1.4190 to 1.4240 | |
0.94 | |
KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
4,4-dimethyl-4,5-dihydro-1,3-oxazole | |
121630 | |
98% | |
4, 4-Dimethyl-2-oxazoline |
97.5 | |
Colorless | |
99.0°C to 100.0°C | |
Authentic | |
C5H9NO | |
MFCD00014507 | |
4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
CC1(C)COC=N1 | |
99.13 | |
99.13 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 250-045-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4,4-Dimethyl-2-oxazoline
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only