missing translation for 'onlineSavingsMsg'
Learn More
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals Product Code.: 11347409

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals

Product Code. 11347409
5 g
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy

Product Code. 11347409

Brand: Thermo Scientific Alfa Aesar H27830.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-46-5
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00020350
InChI Key OSCKRHPYZNTEIO-UHFFFAOYSA-M
Synonym p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acidShow MoreShow Less
PubChem CID 23674498
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 291.28
Grade Indicator
Chemical Name or Material 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10147336 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH27830.06
Lot Number10147336 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27830.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.