missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexan-1-ol, 95%, Thermo Scientific™

Product Code. 10547473
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 932-01-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00101954
InChI Key VUQOIZPFYIVUKD-UHFFFAOYSA-N
PubChem CID 136735
IUPAC Name 4,4-dimethylcyclohexan-1-ol
SMILES CC1(C)CCC(O)CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 4,4-dimethylcyclohexanol, cyclohexanol, 4,4-dimethyl, 4,4-dimethyl-cyclohexanol, cyclohexanol,4,4-dimethyl, 4,4-dimethyl-1-cyclohexanol, 4,4-dimethyl cyclohexan-1-ol
Formula Weight 128.21
Percent Purity 95%
Chemical Name or Material 4, 4-Dimethylcyclohexan-1-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'