missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%, Thermo Scientific™

Product Code. 11348765
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93693-01-7
Molecular Formula C9H13N3S
Molecular Weight (g/mol) 195.28
MDL Number MFCD00041303
InChI Key MXVVZBDSLDIVAW-UHFFFAOYSA-N
Synonym 4-4-ethylphenyl-3-thiosemicarbazide, n-4-ethylphenyl hydrazinecarbothioamide, 3-amino-1-4-ethylphenyl thiourea, 4-ethylphenyl amino hydrazinomethane-1-thione, 1-amino-3-4-ethylphenyl thiourea, 4-4'-ethylphenyl-3-thiosemicarbazide
PubChem CID 3259670
IUPAC Name 1-amino-3-(4-ethylphenyl)thiourea
SMILES CCC1=CC=C(NC(=S)NN)C=C1

Specifications

Melting Point 128°C to 131°C
Quantity 5 g
UN Number UN2811
Beilstein 2804749
Formula Weight 195.29
Percent Purity ≥98%
Chemical Name or Material 4-(4-Ethylphenyl)-3-thiosemicarbazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'