missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)pyridine, 99%, Thermo Scientific Chemicals

Product Code. 11406036
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41538-36-7
Molecular Formula C12H8FNO
Molecular Weight (g/mol) 201.20
MDL Number MFCD02683088
InChI Key WTRWBYGUMQEFFI-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone
PubChem CID 7023019
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
IUPAC Name (4-fluorophenyl)-pyridin-4-ylmethanone
Formula Weight 201.2
Percent Purity 99%
Chemical Name or Material 4-(4-Fluorobenzoyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'