missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)butyric acid, 97%, Thermo Scientific Chemicals

Product Code. 11321969
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-06-0
Molecular Formula C10H11FO2
Molecular Weight (g/mol) 182.19
MDL Number MFCD03788503
InChI Key XVQYBBYOYJXQBF-UHFFFAOYSA-N
Synonym 4-4-fluorophenyl butanoic acid, 4-4-fluorophenyl butyric acid, 4-4-fluoro-phenyl-butyric acid, 4-p-fluorophenyl butyric acid, benzenebutanoic acid, 4-fluoro, 4-4-fluorophenyl butanoicacid, acmc-1awzu, 4-fluorobenzenebutanoic acid, 4-4-fluorophenyl butyricacid, benzenebutanoic acid,4-fluoro
PubChem CID 68524
IUPAC Name 4-(4-fluorophenyl)butanoic acid
SMILES OC(=O)CCCC1=CC=C(F)C=C1

Specifications

Melting Point 44°C to 46°C
Quantity 1 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenyl)butyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'