missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Iodophenyl)morpholine, Thermo Scientific™

Product Code. 10556913
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 87350-77-4
Molecular Formula C10H12INO
Molecular Weight (g/mol) 289.116
InChI Key YUEREUIIUISPFV-UHFFFAOYSA-N
PubChem CID 2795358
IUPAC Name 4-(4-iodophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)I

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-4-iodophenyl morpholine, 4-iodophenylmorpholine, morpholine,4-4-iodophenyl, morpholine, 4-4-iodophenyl, 1-iodo-4-morpholin-4-yl benzene
Formula Weight 289.11
Percent Purity 97%
Chemical Name or Material 4-(4-Iodophenyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'