missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15489155
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871366-38-0
Molecular Formula C20H24BNO3
Molecular Weight (g/mol) 337.226
MDL Number MFCD10567027
InChI Key FEZZNCDZVGVBHZ-UHFFFAOYSA-N
Synonym 4-4-methoxyphenylimino methyl phenylboronic acid pinacol ester, 4-4-methoxyphenyl iminomethylphenylboronic acid pinacol ester, 4-4-methoxyphenyliminomethyl benzeneboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylidene-4-methoxybenzenamine, 1e-n-4-methoxyphenyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb092, 4-4-methoxyphenyl iminomethyl phenyl boronic acid pinacol ester, 1-methoxyphenyliminomethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
PubChem CID 46739070
IUPAC Name N-(4-methoxyphenyl)-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=NC3=CC=C(C=C3)OC

Specifications

Quantity 5 g
Formula Weight 337.22
Chemical Name or Material 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'