Learn More
4,4'-Methylenebis(N,N-dimethylaniline), 98%, Thermo Scientific Chemicals
£26.20 - £288.00
Chemical Identifiers
CAS | 101-61-1 |
---|---|
Molecular Formula | C17H22N2 |
Molecular Weight (g/mol) | 254.37 |
MDL Number | MFCD00008317 |
InChI Key | JNRLEMMIVRBKJE-UHFFFAOYSA-N |
Synonym | 4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone |
PubChem CID | 7567 |
ChEBI | CHEBI:34370 |
IUPAC Name | 4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline |
SMILES | CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031450
|
Thermo Scientific Acros
126740050 |
5 g | Glass bottle |
£26.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10489430
|
Thermo Scientific Acros
126741000 |
100 g | Glass bottle |
£60.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499430
|
Thermo Scientific Acros
126745000 |
500 g | Glass bottle |
£288.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101-61-1 | |
254.37 | |
JNRLEMMIVRBKJE-UHFFFAOYSA-N | |
7567 | |
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline |
C17H22N2 | |
MFCD00008317 | |
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone | |
CHEBI:34370 | |
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C |
Specifications
101-61-1 | |
100.0 | |
Beige-Brown or Gray or White | |
178°C | |
98% | |
H2NC6H4CH2C6H4NH2 | |
13, 239 | |
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone | |
JNRLEMMIVRBKJE-UHFFFAOYSA-N | |
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline | |
7567 | |
254.37 | |
Crystalline Powder, Crystals or Chunks |
99.0 | |
86.0°C to 90.0°C | |
390.0°C | |
Authentic | |
C17H22N2 | |
MFCD00008317 | |
15, 6255 | |
Solubility in water: insoluble. Other solubilities: soluble in benzene,ether,carbon disulfide and,acids,slightly soluble in cold alcohol,more soluble in,hot alcohol | |
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C | |
254.37 | |
CHEBI:34370 | |
98% | |
4, 4'-Methylenebis(N, N-dimethylaniline), 99+% |
Safety and Handling
GHS H Statement:
May cause cancer.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-959-2
missing translation for 'rtecsNumber' : BY5250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only