missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-methylperhydro-1,4-diazepin-1-yl)benzonitrile, Thermo Scientific™
£48.30 - £219.00
Chemical Identifiers
CAS | 166438-80-8 |
---|---|
Molecular Formula | C13H17N3 |
Molecular Weight (g/mol) | 215.3 |
InChI Key | LWRMQRYMWJWCLS-UHFFFAOYSA-N |
Synonym | 4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile |
PubChem CID | 18001106 |
IUPAC Name | 4-(4-methyl-1,4-diazepan-1-yl)benzonitrile |
SMILES | CN1CCCN(CC1)C2=CC=C(C=C2)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10327064
|
Thermo Scientific Maybridge
CC60316DA |
1 g |
£48.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10704097
|
Thermo Scientific Maybridge
CC60316DE |
5 g |
£219.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
166438-80-8 | |
215.3 | |
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile | |
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile |
C13H17N3 | |
LWRMQRYMWJWCLS-UHFFFAOYSA-N | |
18001106 | |
CN1CCCN(CC1)C2=CC=C(C=C2)C#N |
Specifications
166438-80-8 | |
C13H17N3 | |
LWRMQRYMWJWCLS-UHFFFAOYSA-N | |
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile | |
18001106 | |
97% |
97% | |
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile | |
CN1CCCN(CC1)C2=CC=C(C=C2)C#N | |
215.3 | |
215.3 | |
4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile |
Safety and Handling
GHS Signal Word: Danger