missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylphenyl)-3-thiosemicarbazide, 98+%, Thermo Scientific™

Product Code. 11327666
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13278-67-6
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00041302
InChI Key IEAWRKQVDLFINI-UHFFFAOYSA-N
Synonym 4-4-methylphenyl-3-thiosemicarbazide, n-p-tolyl hydrazinecarbothioamide, n-4-methylphenyl hydrazinecarbothioamide, 3-amino-1-4-methylphenyl thiourea, 1-amino-3-4-methylphenyl thiourea, n1-4-methylphenyl hydrazine-1-carbothioamide, hydrazino 4-methylphenyl amino methane-1-thione, acmc-20amvq, 4-p-tolylthiosemicarbazide, 4-p-tolylthiosemicarbazid
PubChem CID 737132
IUPAC Name 1-amino-3-(4-methylphenyl)thiourea
SMILES CC1=CC=C(C=C1)NC(=S)NN

Specifications

Melting Point 135°C to 136°C
Quantity 25 g
UN Number UN2811
Beilstein 879741
Formula Weight 181.26
Percent Purity ≥98%
Chemical Name or Material 4-(4-Methylphenyl)-3-thiosemicarbazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'