missing translation for 'onlineSavingsMsg'
Learn More

{4-[(4-Methylpiperazin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™

Product Code. 10537683
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 622381-65-1
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD07772866
InChI Key ADMFZWUKZBAYIJ-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl phenyl methanol, 4-4-methyl-1-piperazinyl methyl phenyl methanol, 4-4-methylpiperazinyl methyl phenyl methan-1-ol
PubChem CID 7162037
SMILES CN1CCN(CC2=CC=C(CO)C=C2)CC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name {4-[(4-methylpiperazin-1-yl)methyl]phenyl}methanol
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material {4-[(4-Methylpiperazin-1-yl)methyl]phenyl}methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.