Learn More
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde, 97%, Thermo Scientific Chemicals
£161.00 - £551.12
Chemical Identifiers
CAS | 307308-03-8 |
---|---|
Molecular Formula | C8H10N2O |
Molecular Weight (g/mol) | 150.18 |
InChI Key | CSPINDKIWMCRNX-UHFFFAOYSA-N |
Synonym | 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde, pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci, pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci |
PubChem CID | 45083680 |
IUPAC Name | 4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde |
SMILES | C1CCN2C(=C(C=N2)C=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10627040
|
Thermo Scientific Acros
431090010 |
1 g |
£161.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10332020
|
Thermo Scientific Acros
431090050 |
5 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
307308-03-8 | |
150.18 | |
4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde, pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci, pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci | |
4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde |
C8H10N2O | |
CSPINDKIWMCRNX-UHFFFAOYSA-N | |
45083680 | |
C1CCN2C(=C(C=N2)C=O)C1 |
Specifications
307308-03-8 | |
100.0 | |
Authentic | |
Glass bottle | |
4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde, pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci, pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci | |
C1CCN2C(=C(C=N2)C=O)C1 | |
150.18 | |
150.18 | |
Liquid or Low Melting Solid |
96.0 | |
White to Yellow | |
96% min. (GC) | |
C8H10N2O | |
CSPINDKIWMCRNX-UHFFFAOYSA-N | |
4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde | |
45083680 | |
97% | |
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only