missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5',8-Trimethylpsoralen, 97.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Melting Point | 230.0°C to 234.0°C |
Color | White to Beige |
Loss on Drying | 0.5% max. (105°C, 6 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96.5% min. (on dried substance) (HPLC) |
Packaging | Glass bottle |
Quantity | 100 mg |
Merck Index | 15, 9907 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- 4,5',8-Trimethylpsoralen
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'