Learn More
4,5',8-Trimethylpsoralen, 97.5%, Thermo Scientific Chemicals
£40.10 - £248.00
Chemical Identifiers
CAS | 3902-71-4 |
---|---|
Molecular Formula | C14H12O3 |
Molecular Weight (g/mol) | 228.25 |
MDL Number | MFCD00005010 |
InChI Key | FMHHVULEAZTJMA-UHFFFAOYSA-N |
Synonym | trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen |
PubChem CID | 5585 |
ChEBI | CHEBI:28329 |
IUPAC Name | 2,5,9-trimethylfuro[3,2-g]chromen-7-one |
SMILES | CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10502061
|
Thermo Scientific Acros
229881000 |
100 mg |
£40.10
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10510594
|
Thermo Scientific Acros
229880010 |
1 g |
£248.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3902-71-4 | |
228.25 | |
FMHHVULEAZTJMA-UHFFFAOYSA-N | |
5585 | |
2,5,9-trimethylfuro[3,2-g]chromen-7-one |
C14H12O3 | |
MFCD00005010 | |
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
CHEBI:28329 | |
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 |
Specifications
3902-71-4 | |
100.0 | |
White to Beige | |
Authentic | |
Glass bottle | |
MFCD00005010 | |
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
FMHHVULEAZTJMA-UHFFFAOYSA-N | |
2,5,9-trimethylfuro[3,2-g]chromen-7-one | |
5585 | |
228.25 | |
Crystalline Powder |
96.5 | |
230.0°C to 234.0°C | |
0.5% max. (105°C, 6 hrs) | |
96.5% min. (on dried substance) (HPLC) | |
C14H12O3 | |
15, 9907 | |
Solubility in water: insoluble. Other solubilities: sparingly soluble in boiling water,slightly soluble in methanol,ethanol,chloroform | |
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 | |
228.25 | |
CHEBI:28329 | |
97.5% | |
4, 5', 8-Trimethylpsoralen, 97.5% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-459-0
missing translation for 'rtecsNumber' : LV1576000
missing translation for 'tsca' : TSCA
- 4,5',8-Trimethylpsoralen
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only