Learn More
4,5-Diazafluoren-9-one, 98%, Thermo Scientific Chemicals
£90.10 - £328.00
Chemical Identifiers
CAS | 50890-67-0 |
---|---|
Molecular Formula | C11H6N2O |
Molecular Weight (g/mol) | 182.182 |
MDL Number | MFCD00046892 |
InChI Key | PFMTUGNLBQSHQC-UHFFFAOYSA-N |
Synonym | 4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one |
PubChem CID | 342157 |
SMILES | C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492448
|
Thermo Scientific Alfa Aesar
H56028.03 |
1 g |
£90.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402458
|
Thermo Scientific Alfa Aesar
H56028.06 |
5 g |
£328.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
Chemical Identifiers
50890-67-0 | |
182.182 | |
PFMTUGNLBQSHQC-UHFFFAOYSA-N | |
342157 |
C11H6N2O | |
MFCD00046892 | |
4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one | |
C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1 |
Specifications
50890-67-0 | |
C11H6N2O | |
4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one | |
PFMTUGNLBQSHQC-UHFFFAOYSA-N | |
182.182 | |
182.18 | |
4,5-Diazafluoren-9-one |
214°C to 217°C | |
MFCD00046892 | |
Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes. | |
C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1 | |
342157 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only