missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4',5'-Dibromofluorescein, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Specifications
Melting Point | 270.0°C to 273.0°C |
Loss on Drying | 5% max. |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 5 g |
Beilstein | 19,V,6,462 |
Merck Index | 15,3027 |
Solubility Information | (1% in hot 0.5 N NH4OH) Clear yellow to orange |
Formula Weight | 490.11 |
Chemical Name or Material | 4',5'-Dibromofluorescein |
Safety and Handling

Product Identifier
- 4',5'-Dibromofluorescein
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4',5'-Dibromofluorescein, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction