missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifica
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 255.1 |
Percent Purity | 97% |
Chemical Name or Material | 4, 5-Dichloro-2-(4-methylphenyl)-2, 3-dihydropyridazin-3-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'