missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitrotoluene, 98%, Thermo Scientific Chemicals

Product Code. 15459468
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127371-50-0
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD04973772
InChI Key DJEBTFSOEQWELL-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-nitrotoluene, 2-nitro-4,5-difluorotoluene, 4,5-difluoro-2-methylnitrobenzene, 4,5-difluoro-1-methyl-2-nitrobenzene, benzene,1,2-difluoro-4-methyl-5-nitro, pubchem18518, acmc-1c6zu, 1-nitro-3,4-difluoro-6-methylbenzene, 1,2-difluoro-4-methyl-5-nitro-benzene, benzene, 1,2-difluoro-4-methyl-5-nitro
PubChem CID 2756252
IUPAC Name 1,2-difluoro-4-methyl-5-nitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])F)F

Specifications

Color Colorless to Yellow
Quantity 1 g
UN Number UN2810
Formula Weight 173.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4,5-Difluoro-2-nitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'