missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, is used to improve the visualization of trichothecenes on developed TLC plates. It serves as an intermediate in the production of dyes, pigments, chelating reagents, sulfonic and sulfinic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | White |
Quantity | 5 g |
Beilstein | 3647189 |
Merck Index | 14,2241 |
Solubility Information | Soluble in water. |
Formula Weight | 400.29 (364.26 Anhydrous) |
Grade | ACS |
Physical Form | Powder |
Chemical Name or Material | 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'