missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific Chemicals
£58.17 - £252.00
Chemical Identifiers
CAS | 705-24-8 |
---|---|
Molecular Formula | C5HCl2F3N2 |
Molecular Weight (g/mol) | 216.97 |
MDL Number | MFCD08436597 |
InChI Key | QFWVAJQVYBRTCL-UHFFFAOYSA-N |
Synonym | 4,6-dichloro-2-trifluoromethyl pyrimidine, 4,6-dichloro-2-trifluoromethyl-pyrimidine, pyrimidine, 4,6-dichloro-2-trifluoromethyl, pubchem19441, ksc496e6h, 4 pound not6-dichloro-2-trifluoromethylpyrimidine |
PubChem CID | 15713197 |
IUPAC Name | 4,6-dichloro-2-(trifluoromethyl)pyrimidine |
SMILES | FC(F)(F)C1=NC(Cl)=CC(Cl)=N1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
705-24-8 | |
216.97 | |
QFWVAJQVYBRTCL-UHFFFAOYSA-N | |
15713197 | |
FC(F)(F)C1=NC(Cl)=CC(Cl)=N1 |
C5HCl2F3N2 | |
MFCD08436597 | |
4,6-dichloro-2-trifluoromethyl pyrimidine, 4,6-dichloro-2-trifluoromethyl-pyrimidine, pyrimidine, 4,6-dichloro-2-trifluoromethyl, pubchem19441, ksc496e6h, 4 pound not6-dichloro-2-trifluoromethylpyrimidine | |
4,6-dichloro-2-(trifluoromethyl)pyrimidine |
Specifications
705-24-8 | |
96% min. (GC) | |
C5HCl2F3N2 | |
4,6-dichloro-2-trifluoromethyl pyrimidine, 4,6-dichloro-2-trifluoromethyl-pyrimidine, pyrimidine, 4,6-dichloro-2-trifluoromethyl, pubchem19441, ksc496e6h, 4 pound not6-dichloro-2-trifluoromethylpyrimidine | |
FC(F)(F)C1=NC(Cl)=CC(Cl)=N1 | |
216.97 | |
216.98 | |
4, 6-Dichloro-2-(trifluoromethyl)pyrimidine |
50°C to 52°C (5.3 mbar) | |
Glass Bottle | |
MFCD08436597 | |
QFWVAJQVYBRTCL-UHFFFAOYSA-N | |
4,6-dichloro-2-(trifluoromethyl)pyrimidine | |
15713197 | |
97% |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4,6-Dichloro-2-(trifluoromethyl)pyrimidine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only