missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichlororesorcinol, 97%, Thermo Scientific™

Product Code. 11424507
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,6-Dichlororesorcinol is a resorcinol (sc-203371) derivative, that acts as an antiseptic agent. It is also used to prepare HMGCR inhibitors. It is used as a pharmaceutical and organic intermediate.

Solubility
Solub
TRUSTED_SUSTAINABILITY

Specifications

CAS 137-19-9
Melting Point 113°C to 116°C
Boiling Point 253°C to 255°C
Molecular Formula C6H4Cl2O2
MDL Number MFCD00002274
Quantity 25 g
Synonym 4,6-dichlororesorcinol, 1,3-benzenediol, 4,6-dichloro, 4,6-dichloro-1,3-benzenediol, 4,6-dichloro-1,3-dihydroxybenzene, resorcinol, 4,6-dichloro, 1,5-dichloro-2,4-dihydroxy-benzene, 4,3-dihydroxybenzene, 3,4-dichlororesorcinol, resorcinol,6-dichloro, acmc-209caz
Solubility Information Soluble in water.
InChI Key GRLQBYQELUWBIO-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1O)Cl)Cl)O
IUPAC Name 4,6-dichlorobenzene-1,3-diol
Molecular Weight (g/mol) 178.996
PubChem CID 67299
Formula Weight 179
Percent Purity 97%
Chemical Name or Material 4,6-Dichlororesorcinol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'