missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Difluoroindole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Sensitivity | Air sensitive |
Synonym | 4,6-difluoroindole, 1h-indole, 4,6-difluoro, 4,6-difluoro-indole, pubchem7179, acmc-209f4a, 1h-indole,4,6-difluoro, 1h-indole,4,6-difluoro-9ci, 4,6-difluoroindole 153.13, 4,6-difluoro-1h-indole; 1h-indole,4,6-difluoro-9ci |
Solubility Information | Sparingly Soluble (0.062g/L) (25°C),Calc. |
Formula Weight | 153.13 |
Percent Purity | 97% |
Chemical Name or Material | 4,6-Difluoroindole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'