missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™

Product Code. 15416187
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-47-9
Molecular Formula C5H3F3N2O2
Molecular Weight (g/mol) 180.09
MDL Number MFCD02678234
InChI Key AMGBKPNVGVAFEN-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidine-4,6-diol, 2-trifluoromethyl-pyrimidine-4,6-diol, 2-trifluoromethylpyrimidine-4,6-diol, 4,6-dihydroxy-2-trifluoromethylpyrimidine, 4 1h-pyrimidinone,6-hydroxy-2-trifluoromethyl, 4,6-dihydroxy-2-trifluoromethyl pyrimidine, 6-hydroxy-2-trifluoromethyl-3h-pyrimidin-4-one, 4-hydroxy-2-trifluoromethyl-1h-pyrimidin-6-one, 6-hydroxy-2-trifluoromethyl-4 1h-pyrimidinone, 6-hydroxy-2-trifluoromethyl-1h-pyrimidin-4-one
PubChem CID 243103
IUPAC Name 4-hydroxy-2-(trifluoromethyl)-1H-pyrimidin-6-one
SMILES OC1=CC(=O)NC(=N1)C(F)(F)F

Specifications

Melting Point 254°C to 256°C
Quantity 1 g
Formula Weight 180.08
Percent Purity 97%
Chemical Name or Material 4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'