missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-5-nitropyrimidine, 95%, Thermo Scientific™

Product Code. p-112462 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10251383

Brand: Thermo Scientific Acros 305410050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2164-83-2
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006110
InChI Key ABTLZAVJDRUDNG-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-4,6-diol, 4,6-dihydroxy-5-nitropyrimdine, 5-nitro-4,6-pyrimidinediol, 4 1h-pyrimidinone, 6-hydroxy-5-nitro, 6-hydroxy-5-nitro-3h-pyrimidin-4-one, 6-hydroxy-5-nitro-4 1h-pyrimidinone, 5-nitro-4,6-dihydroxypyrimidine, 4-hydroxy-5-nitro-1h-pyrimidin-6-one, 4 3h-pyrimidinone, 6-hydroxy-5-nitro
PubChem CID 75105
IUPAC Name 6-hydroxy-5-nitro-3,4-dihydropyrimidin-4-one
SMILES OC1=C(C(=O)NC=N1)[N+]([O-])=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.09
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.