missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-(methylsulfonyl)pyrimidine, 98%, Thermo Scientific™

Product Code. 15491049
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35144-22-0
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD03788286
InChI Key ZHPSNGCLCHWTRG-UHFFFAOYSA-N
Synonym 4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d
PubChem CID 2772382
IUPAC Name 4,6-dimethyl-2-methylsulfonylpyrimidine
SMILES CC1=CC(=NC(=N1)S(=O)(=O)C)C

Specifications

Melting Point 80°C to 85°C (literature)
Color Red to Brown
Quantity 5 g
Beilstein 778406
Formula Weight 186.23
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4,6-Dimethyl-2-(methylsulfonyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'