missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4,6-O-Ethylidene-D-glucopyranose, 90%

Product Code. 15407999
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15407999

Brand: Thermo Scientific Chemicals J66583.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13224-99-2 | C8H14O6 | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13224-99-2
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00006820
InChI Key VZPBLPQAMPVTFO-UHFFFAOYNA-N
Synonym 4,6-o-ethylidene-a-d-glucose
PubChem CID 21581900
IUPAC Name (2R,4aR,6S,7R,8R,8aS)-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-6,7,8-triol
SMILES CC1OCC2OC(O)C(O)C(O)C2O1

Specifications

Color White
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 206.19
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 4,6-O-Ethylidene-D-glucopyranose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.