missing translation for 'onlineSavingsMsg'
Learn More
4,6-O-Isopropylidene-D-glucal, 97%, Thermo Scientific Chemicals Product Code.: 15466698

4,6-O-Isopropylidene-D-glucal, 97%, Thermo Scientific Chemicals

Product Code. 15466698
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15466698

Brand: Thermo Scientific Alfa Aesar H60221.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51450-36-3
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD22988998
InChI Key GAQDIYMHBQNXLE-PRJMDXOYSA-N
Synonym 4,6-o-isopropylidene-d-glucal, 4-o,6-o-isopropylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,2-didehydro-1,2-dideoxy-4-o,6-o-isopropylidene-beta-d-glucopyranose, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 11052343
IUPAC Name (4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES CC1(OCC2C(O1)C(C=CO2)O)C

Specifications

Quantity 1 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4,6-O-Isopropylidene-D-glucal

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA15Z020 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60221.03
Lot NumberQ14G019 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60221.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,6-O-Isopropylidene-D-glucal, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.