missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
5 g
Description
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.
Solubility
Soluble in benzene and alcohol. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.
Solubility
Soluble in benzene and alcohol. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 193°C to 195°C |
Quantity | 1 g |
Solubility Information | Soluble in benzene and alcohol. Slightly soluble in water. |
Formula Weight | 208.26 |
Percent Purity | 98% |
Chemical Name or Material | 4,7-Dimethyl-1,10-phenanthroline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'