missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dimethyl-4-octanol, 99%, Thermo Scientific™

Product Code. 11459686
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19781-13-6
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00046603
InChI Key OTISWHNJCUMBFI-UHFFFAOYSA-N
PubChem CID 89224
IUPAC Name 4,7-dimethyloctan-4-ol
SMILES CCCC(C)(CCC(C)C)O

Specifications

Quantity 5 g
Synonym 4,7-dimethyl-4-octanol, 4-octanol, 4,7-dimethyl, 2,5-dimethyl-5-octanol, acmc-20amr9, 2.5-dimethyloctanol-5, 4-octanol,4,7-dimethyl
Formula Weight 158.29
Percent Purity 99%
Chemical Name or Material 4,7-Dimethyl-4-octanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'