missing translation for 'onlineSavingsMsg'
Learn More
4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid, 95%, Thermo Scientific™ Product Code.: 10252832

4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10252832
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10252832

Brand: Thermo Scientific Maybridge KM01115DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-58-3
Molecular Formula C8H8N4O2
Molecular Weight (g/mol) 192.178
MDL Number MFCD00067873
InChI Key JFCXJLMOHOXSTO-UHFFFAOYSA-N
Synonym 4,7-dimethylpyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, 4,7-dimethylpyrazolo 3,2-c 1,2,4 triazine-3-carboxylic acid, 3-carboxy-4,7-dimethylpyrazolo 5,1-c 1,2,4 triazine, maybridge1_004683, 4,7-dimethylpyrazolo 5,1-c 1,2,4-triazine-3-carboxylic acid, 4,7-dimethylpyrazolo 5,1-c triazine-3-carboxylic acid, 4,7dimethyl-pyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, 4,7-dimethyl, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylicacid, 4,7-dimethyl, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylicacid,4,7-dimethyl
PubChem CID 2778429
IUPAC Name 4,7-dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
SMILES CC1=NN2C(=C(N=NC2=C1)C(=O)O)C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.18
Percent Purity 95%
Chemical Name or Material 4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.