missing translation for 'onlineSavingsMsg'
Learn More

4,8-Dihydroxyquinoline-2-carboxylic acid, 96%, Thermo Scientific Chemicals

Product Code. 10754221
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-00-7
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.17
MDL Number MFCD00006754
InChI Key FBZONXHGGPHHIY-UHFFFAOYSA-N
Synonym xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
PubChem CID 5699
ChEBI CHEBI:10072
SMILES OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 297°C to 298°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10266
IUPAC Name 8-hydroxy-4-oxo-1H-quinoline-2-carboxylic acid
Formula Weight 205.17
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4, 8-Dihydroxyquinoline-2-carboxylic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 4,8-Dihydroxyquinoline-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.