missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-3-nitrobenzoic acid, 97+%, Thermo Scientific™

Product Code. 11424163
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1539-06-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00014703
InChI Key BRQIMWBIZLRLSV-UHFFFAOYSA-N
Synonym 4-acetamido-3-nitrobenzoic acid, 4-acetylamino-3-nitrobenzoic acid, benzoic acid, 4-acetylamino-3-nitro, unii-9w2250fzkc, 4-acetamino-3-nitrobenzoic acid, 4-acetamido-3-nitro-benzoic acid, 4-acetylamino-3-nitro-benzoic acid, pubchem16827, acmc-1bqk5
PubChem CID 73758
SMILES CC(=O)NC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 223°C to 228°C
Quantity 25 g
Beilstein 2217819
Formula Weight 224.17
Percent Purity ≥97%
Chemical Name or Material 4-Acetamido-3-nitrobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'