missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-9-fluorenone, 99%, Thermo Scientific™

Product Code. 11480537
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42135-35-3
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.258
MDL Number MFCD00010782
InChI Key BDYZWJVQPFNPBV-UHFFFAOYSA-N
Synonym 4-acetamido-9-fluorenone, n-9-oxo-9h-fluoren-4-yl acetamide, n-9-oxofluoren-4-yl acetamide, acmc-1asup, 4-acetylamino-9-fluorenone, n-9-oxo-4-fluorenyl acetamide, n-9-oxo-9h-fluoren-4-yl acetamide #, n-9-oxidanylidenefluoren-4-yl ethanamide
PubChem CID 235270
IUPAC Name N-(9-oxofluoren-4-yl)acetamide
SMILES CC(=O)NC1=CC=CC2=C1C3=CC=CC=C3C2=O

Specifications

Melting Point 262°C to 264°C
Quantity 5 g
Formula Weight 237.26
Percent Purity 99%
Chemical Name or Material 4-Acetamido-9-fluorenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'