missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10623583
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-85-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00003380
InChI Key SKLUWKYNZNXSLX-UHFFFAOYSA-N
Synonym 4-acetamidobenzaldehyde, n-4-formylphenyl acetamide, 4-acetylaminobenzaldehyde, p-formylacetanilide, acetamide, n-4-formylphenyl, micotiazone, 4-formylacetanilide, 4'-formylacetanilide, p-acetamidobenzaldehyde, p-acetaminobenzaldehyde
PubChem CID 73942
IUPAC Name N-(4-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 154°C to 158°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula OHCC6H4NHCOCH3
Quantity 25 g
Beilstein 14,IV,71
Solubility Information Solubility in water: slightly soluble
Formula Weight 163.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidobenzaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetamidobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'