missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzenesulfonyl azide, 97%

Product Code. 15362317 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15362317

Brand: Thermo Scientific Acros 443800250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2158-14-7 | C8H8N4O3S | 240.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2158-14-7
Molecular Formula C8H8N4O3S
Molecular Weight (g/mol) 240.24
MDL Number MFCD00029626
InChI Key NTMHWRHEGDRTPD-UHFFFAOYSA-N
Synonym 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
PubChem CID 5129185
IUPAC Name N-(4-azidosulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]

Specifications

Melting Point 107.0°C to 111.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 240.24
Percent Purity 97%
Chemical Name or Material 4-Acetamidobenzenesulfonyl azide
compliance-icons
Product Identifier
  • 4-Acetamidobenzenesulfonyl azide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.