missing translation for 'onlineSavingsMsg'
Learn More
4-Acetoxy-2-azetidinone, 97%, Thermo Scientific Chemicals Product Code.: 15535926

4-Acetoxy-2-azetidinone, 97%, Thermo Scientific Chemicals

Product Code. 15535926
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15535926

Brand: Thermo Scientific Acros 456640010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28562-53-0
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00010593
InChI Key OEYMQQDJCUHKQS-UHFFFAOYNA-N
Synonym 4-acetoxy-2-azetidinone, 2-azetidinone, 4-acetyloxy, 4-acetoxyazetidinone, 4-oxoazetidin-2-yl acetate, 2-oxoazetidinium 4-acetate, 2-azetidinone, 4-acetyloxy-, 4r, 4-acetoxy-azetidinone, acmc-20bsq9, 4-acetoxyazetidin-2-one, 4-acetoxyazetidine-2-one
PubChem CID 119981
IUPAC Name (4-oxoazetidin-2-yl) acetate
SMILES CC(=O)OC1CC(=O)N1

Specifications

Melting Point 35.0°C to 38.0°C
Elemental Analysis 44.65 to 48.37% (C) 10.41 to 11.28% (N)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.12
Percent Purity 97%
Chemical Name or Material 4-Acetoxy-2-azetidinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number344224 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number456640050,456640010
Lot Number344081 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number456640050,456640010
compliance-icons compliance-icons
Product Identifier
  • 4-Acetoxy-2-azetidinone, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Acetoxy-2-azetidinone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.